Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r26
Entrez Gene ID
292531
Gene Name
vomeronasal 2 receptor, 26
Gene Aliases
Gm1961
Chromosome Location
Chr.1: 63535051 - 63571794 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099471.1 NP_001092941.1 1 - 2 197 123

View Full Product Details