Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ces1a
Entrez Gene ID
679817
Gene Name
carboxylesterase 1A
Gene Aliases
-
Chromosome Location
Chr.19: 15339152 - 15366079 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190375.1 NP_001177304.1 2 - 3 255 58

View Full Product Details