Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vom2r11
Entrez Gene ID
292417
Gene Name
vomeronasal 2 receptor, 11
Gene Aliases
RGD1563778
Chromosome Location
Chr.1: 86604426 - 86624214 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099470.1 NP_001092940.1 2 - 3 512 93

View Full Product Details