The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105785.1 | NP_001099255.1 | 1 - 2 | 238 | 72 |
XM_006246460.2 | XP_006246522.1 | 1 - 2 | 685 | 72 | |
GenBank mRNA | BC167058.1 | - | 1 - 2 | 294 | 72 |