Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krt25
Entrez Gene ID
303519
Gene Name
keratin 25, type I
Gene Aliases
Ka38, Krt25A
Chromosome Location
Chr.10: 87225446 - 87232723 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008822.1 NP_001008822.1 7 - 8 1231 72

View Full Product Details