Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd31
Entrez Gene ID
100359720
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.2: 27571133 - 27743163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760712.1 XP_008758934.1 14 - 15 2150 162

View Full Product Details