Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plcxd2
Entrez Gene ID
363781
Gene Name
phosphatidylinositol-specific phospholipase C, X domain containing 2
Gene Aliases
RGD1563504
Chromosome Location
Chr.11: 57207656 - 57260001 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134481.1 NP_001127953.1 3 - 4 1425 63

View Full Product Details