The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033958.2 | NP_001029130.2 | - | 291 | 109 |
NM_001033959.2 | NP_001029131.1 | - | 291 | 109 | |
NM_147215.2 | NP_671748.2 | - | 291 | 109 | |
GenBank mRNA | AB039825.1 | - | - | 272 | 109 |
BC086942.1 | - | - | 261 | 109 | |
J00738.1 | - | - | 291 | 109 | |
X14552.1 | - | - | 287 | 109 |