Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Svs6
Entrez Gene ID
362267
Gene Name
seminal vesicle secretory protein 6
Gene Aliases
Svp6
Chromosome Location
Chr.3: 160695888 - 160697559 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102403.1 NP_001095873.1 1 - 2 99 152

View Full Product Details