Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rac3
Entrez Gene ID
688319
Gene Name
ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3)
Gene Aliases
-
Chromosome Location
Chr.10: 109903920 - 109906596 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008768496.1 XP_008766718.1 5 - 6 1033 156

View Full Product Details