Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cntd1
Entrez Gene ID
498001
Gene Name
cyclin N-terminal domain containing 1
Gene Aliases
RGD1564542
Chromosome Location
Chr.10: 89199988 - 89210369 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197217.1 NP_001184146.1 2 - 3 356 82

View Full Product Details