Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C4b
Entrez Gene ID
406161
Gene Name
complement component 4B (Chido blood group)
Gene Aliases
C4-2, C4l
Chromosome Location
Chr.20: 4302344 - 4316752 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002805.2 NP_001002805.1 13 - 14 1740 96

View Full Product Details