The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001103359.2 | NP_001096829.2 | 10 - 11 | 1191 | 130 |
GenBank mRNA | BC128711.1 | - | 10 - 11 | 1173 | 130 |
U10697.1 | - | 10 - 11 | 1171 | 130 | |
X81825.1 | - | 10 - 11 | 1170 | 130 |