Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxi2
Entrez Gene ID
246073
Gene Name
forkhead box I2
Gene Aliases
Fkhl5, Foxf1
Chromosome Location
Chr.1: 207653703 - 207656831 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_341949.4 XP_341950.2 1 - 2 1367 142

View Full Product Details