Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cfhr1
Entrez Gene ID
289057
Gene Name
complement factor H-related 1
Gene Aliases
Cfhl1
Chromosome Location
Chr.13: 56862414 - 56890208 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044227.1 NP_001037692.1 5 - 5 746 146
XM_006249941.1 XP_006250003.1 6 - 6 857 146
GenBank mRNA BC088173.1 - 5 - 5 746 146
FQ218705.1 - 5 - 5 764 146
FQ219461.1 - 5 - 5 767 146

View Full Product Details