The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021598.2 | NP_067609.1 | - | 207 | 139 |
GenBank mRNA | FQ229669.1 | - | - | 226 | 139 |
U67911.1 | - | - | 208 | 139 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_003751469.3 | XP_003751517.1 | 2 - 3 | 268 | 139 |