The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173304.2 | NP_775426.1 | 9 - 9 | 1435 | 75 |
GenBank mRNA | BC088097.1 | - | 9 - 9 | 1437 | 75 |
J02869.1 | - | 9 - 9 | 1435 | 75 | |
M22329.1 | - | 9 - 9 | 1369 | 75 |