Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xkr7
Entrez Gene ID
311549
Gene Name
XK, Kell blood group complex subunit-related family, member 7
Gene Aliases
-
Chromosome Location
Chr.3: 148508619 - 148537904 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012092.1 NP_001012092.1 3 - 3 1859 94
XM_008762334.1 XP_008760556.1 4 - 4 3583 94
GenBank mRNA AY534261.1 - 3 - 3 1859 94

View Full Product Details