The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080905.2 | NP_543181.2 | 2 - 2 | 1914 | 130 |
XM_006255211.2 | XP_006255273.1 | 2 - 2 | 6515 | 130 | |
XM_006255213.2 | XP_006255275.1 | 2 - 2 | 2254 | 130 | |
XM_008772345.1 | XP_008770567.1 | 3 - 3 | 1955 | 130 | |
GenBank mRNA | BQ780107.1 | - | 1 - 1 | 359 | 130 |