Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spry3
Entrez Gene ID
498159
Gene Name
sprouty homolog 3 (Drosophila)
Gene Aliases
RGD1562172
Chromosome Location
Chr.12: 18931374 - 18940622 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109063.1 NP_001102533.1 2 - 2 443 106
XM_008769065.1 XP_008767287.1 2 - 2 1817 106
XM_008769066.1 XP_008767288.1 3 - 3 738 106
XM_008769067.1 XP_008767289.1 3 - 3 690 106
XM_008769068.1 XP_008767290.1 2 - 2 406 106
XM_008769069.1 XP_008767291.1 2 - 2 536 106
GenBank mRNA BC168971.1 - 2 - 2 436 106

View Full Product Details