The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133603.2 | NP_598287.1 | 2 - 2 | 1526 | 154 |
XM_006248023.2 | XP_006248085.1 | 2 - 2 | 1952 | 154 | |
XM_008768554.1 | XP_008766776.1 | 2 - 2 | 1762 | 154 | |
GenBank mRNA | BC089778.1 | - | 2 - 2 | 1526 | 154 |