Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pard6g
Entrez Gene ID
307237
Gene Name
par-6 family cell polarity regulator gamma
Gene Aliases
-
Chromosome Location
Chr.18: 76559877 - 76627843 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100973.1 NP_001094443.1 3 - 3 927 176
GenBank mRNA EF210570.1 - 3 - 3 927 176

View Full Product Details