Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Angel1
Entrez Gene ID
362765
Gene Name
angel homolog 1 (Drosophila)
Gene Aliases
RGD1306238
Chromosome Location
Chr.6: 110637881 - 110681407 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006240392.2 XP_006240454.1 11 - 11 109
XM_008764815.1 XP_008763037.1 10 - 10 109

View Full Product Details