Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Irf2bpl
Entrez Gene ID
314329
Gene Name
interferon regulatory factor 2 binding protein-like
Gene Aliases
Eap1, Pqcp, RGD1310994
Chromosome Location
Chr.6: 110901940 - 110904288 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012470.1 NP_001012488.1 1 - 1 2248 155
GenBank mRNA BC098759.1 - 1 - 1 949 155

View Full Product Details