Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fos
- Entrez Gene ID
-
314322
- Gene Name
- FBJ osteosarcoma oncogene
- Gene Aliases
-
c-fos
- Chromosome Location
-
Chr.6: 109300433 - 109303299 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X06769.1 |
- |
- |
1894 |
181 |
View Full Product Details