The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105935.1 | NP_001099405.1 | 12 - 12 | 2636 | 167 |
XM_006249493.2 | XP_006249555.1 | 11 - 11 | 3316 | 167 |