The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105928.1 | NP_001099398.1 | 4 - 4 | 2378 | 106 |
XM_006249332.2 | XP_006249394.1 | 3 - 3 | 2861 | 106 |