The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105917.1 | NP_001099387.2 | 14 - 14 | 1481 | 88 |
XM_006249120.2 | XP_006249182.1 | 14 - 14 | 1562 | 88 |