Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom1r7
Entrez Gene ID
494259
Gene Name
vomeronasal 1 receptor 7
Gene Aliases
V1re3
Chromosome Location
Chr.1: 60087495 - 60088406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008907.1 NP_001008907.1 1 - 1 506 108
GenBank mRNA AY510304.1 - 1 - 1 506 108

View Full Product Details