The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105948.1 | NP_001099418.1 | 7 - 7 | 2853 | 180 |
XM_008769428.1 | XP_008767650.1 | 7 - 7 | 3091 | 180 | |
XM_008769429.1 | XP_008767651.1 | 8 - 8 | 3663 | 180 |