Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Cntnap5a
Entrez Gene ID
297832
Gene Name
contactin associated protein-like 5A
Gene Aliases
Caspr5-1, RGD1559812
Chromosome Location
Chr.13: 6510703 - 6608827 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047865.1 NP_001041330.1 - 958 114

View Full Product Details