The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_171988.2 | NP_741985.1 | 4 - 4 | 4984 | 150 |
XM_006234983.2 | XP_006235045.2 | 4 - 4 | 5102 | 150 | |
XM_006234985.2 | XP_006235047.2 | 5 - 5 | 4934 | 150 | |
XM_006234987.2 | XP_006235049.2 | 4 - 4 | 4844 | 150 |