The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019129.3 | NP_062002.1 | 2 - 2 | 385 | 95 |
GenBank mRNA | BP465191.1 | - | 1 - 1 | 337 | 95 |
BP502578.1 | - | 2 - 2 | 407 | 95 | |
C07095.1 | - | 1 - 1 | 184 | 95 |