The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099507.1 | NP_001092977.1 | 6 - 6 | 2416 | 139 |
GenBank mRNA | AF053988.1 | - | 1 - 1 | 377 | 139 |