The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172329.1 | NP_758832.1 | 3 - 3 | 2041 | 127 |
XM_006244179.2 | XP_006244241.1 | 3 - 3 | 2847 | 127 | |
GenBank mRNA | AF458780.1 | - | 3 - 3 | 2041 | 127 |
BC091115.1 | - | 3 - 3 | 2055 | 127 |