The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002288.1 | NP_001002288.1 | 1 - 1 | 856 | 132 |
XM_006230870.2 | XP_006230932.1 | 4 - 4 | 1422 | 132 | |
XM_006230872.2 | XP_006230934.1 | 2 - 2 | 1220 | 132 | |
XM_008760163.1 | XP_008758385.1 | 3 - 3 | 1318 | 132 |