The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007556.1 | NP_001007557.1 | 3 - 4 | 801 | 126 |
GenBank mRNA | AY758558.1 | - | 3 - 4 | 801 | 126 |