Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr77
Entrez Gene ID
310769
Gene Name
WD repeat domain 77
Gene Aliases
RGD1310479
Chromosome Location
Chr.2: 208577292 - 208587263 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY321348.1 - 7 - 8 676 155

View Full Product Details