Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt76
Entrez Gene ID
407757
Gene Name
keratin 76, type II
Gene Aliases
Kb9
Chromosome Location
Chr.7: 143489160 - 143497108 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008805.1 NP_001008805.1 7 - 8 1418 105

View Full Product Details