Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kb23
Entrez Gene ID
407759
Gene Name
type II keratin 23
Gene Aliases
-
Chromosome Location
Chr.7: 143135074 - 143141634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008813.1 NP_001008813.1 7 - 8 1248 158

View Full Product Details