Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-A3
Entrez Gene ID
309627
Gene Name
RT1 class I, locus A3
Gene Aliases
RT1-A, RT1.A3
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008830.1 NP_001008830.1 3 - 4 335 124

View Full Product Details