The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008847.2 | NP_001008847.1 | 3 - 4 | 664 | 126 |
GenBank mRNA | AY436750.1 | - | 3 - 4 | 609 | 126 |
AY626210.1 | - | 3 - 4 | 609 | 126 | |
AY626211.1 | - | 3 - 4 | 609 | 126 | |
AY626213.1 | - | 3 - 4 | 609 | 126 | |
AY626215.1 | - | 3 - 4 | 609 | 126 | |
AY626216.1 | - | 3 - 4 | 609 | 126 | |
AY626217.1 | - | 3 - 4 | 609 | 126 | |
AY701539.1 | - | 3 - 4 | 614 | 126 | |
BC098677.1 | - | 3 - 4 | 630 | 126 | |
FQ220763.1 | - | 3 - 4 | 652 | 126 | |
FQ220966.1 | - | 3 - 4 | 657 | 126 | |
FQ226848.1 | - | 3 - 4 | 655 | 126 | |
FQ227693.1 | - | 3 - 4 | 656 | 127 | |
FQ230207.1 | - | 3 - 4 | 655 | 126 | |
FQ232717.1 | - | 3 - 4 | 652 | 126 | |
FQ232757.1 | - | 2 - 3 | 419 | 126 | |
FQ233042.1 | - | 3 - 4 | 655 | 126 | |
FQ233454.1 | - | 3 - 4 | 647 | 126 | |
FQ233943.1 | - | 3 - 4 | 655 | 126 | |
FQ234684.1 | - | 3 - 4 | 658 | 127 | |
FQ235025.1 | - | 3 - 4 | 647 | 126 | |
KC222932.1 | - | 3 - 4 | 609 | 126 | |
KC222933.1 | - | 3 - 4 | 609 | 126 | |
KC222934.1 | - | 3 - 4 | 609 | 126 | |
KC222935.1 | - | 3 - 4 | 609 | 126 |