Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-O1
Entrez Gene ID
24751
Gene Name
RT1 class Ib, locus O1
Gene Aliases
Qlikea, RT1-O
Chromosome Location
Chr.20: 3160535 - 3162939 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008856.1 NP_001008856.1 1 - 2 71 148

View Full Product Details