Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cirh1a
Entrez Gene ID
291987
Gene Name
cirrhosis, autosomal recessive 1A (cirhin)
Gene Aliases
Cirhin, Naic, Teg-292, Tex292
Chromosome Location
Chr.19: 39087995 - 39116124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009640.1 NP_001009640.1 4 - 5 477 144
XM_006255562.1 XP_006255624.1 4 - 5 523 144
XM_006255563.2 XP_006255625.1 4 - 5 686 144
XM_008772488.1 XP_008770710.1 4 - 5 476 144
GenBank mRNA BC088461.1 - 4 - 5 477 144

View Full Product Details