Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pepd
Entrez Gene ID
292808
Gene Name
peptidase D
Gene Aliases
-
Chromosome Location
Chr.1: 90820670 - 91285128 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009641.1 NP_001009641.1 7 - 8 562 168
GenBank mRNA BC088440.1 - 7 - 8 562 168

View Full Product Details