The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009682.1 | NP_001009682.1 | 3 - 4 | 722 | 123 |
XM_008769327.1 | XP_008767549.1 | 3 - 4 | 896 | 123 | |
GenBank mRNA | AY237116.1 | - | 3 - 4 | 722 | 123 |