Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vom1r33
Entrez Gene ID
292580
Gene Name
vomeronasal 1 receptor 33
Gene Aliases
V1rd15, V1rd22
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008942.1 NP_001008942.1 1 - 1 852 94
GenBank mRNA AY510340.1 - 1 - 1 852 94

Gene Symbol
Vom1r39
Entrez Gene ID
100312670
Gene Name
vomeronasal 1 receptor 39
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167551.1 NP_001161023.1 1 - 1 852 94

View Full Product Details