Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tbc1d22a
Entrez Gene ID
689443
Gene Name
TBC1 domain family, member 22a
Gene Aliases
RGD1306588
Chromosome Location
Chr.7: 127080155 - 127225839 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006242171.2 XP_006242233.1 1 - 1 1327 100
XM_008765824.1 XP_008764046.1 1 - 1 1325 100

View Full Product Details