The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007635.2 | NP_001007636.2 | 25 - 26 | 3701 | 152 |
XM_006232134.2 | XP_006232196.1 | 27 - 28 | 4043 | 152 | |
GenBank mRNA | AY631396.1 | - | 25 - 26 | 3701 | 152 |
AY631398.1 | - | 17 - 18 | 2640 | 152 | |
AY631399.1 | - | 25 - 26 | 3466 | 152 | |
AY631400.1 | - | 21 - 22 | 2840 | 152 |