The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008555.1 | NP_001008555.1 | 2 - 3 | 798 | 97 |
GenBank mRNA | AY704415.1 | - | 2 - 3 | 798 | 97 |
AY825923.1 | - | 2 - 3 | 798 | 97 |